5-Bromo-N,N-diethyl-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

051775-500MG 051775-2.5G
101896-992EA 384.62 USD
101896-992 101896-994
5-Bromo-N,N-diethyl-4-methyl-2-pyridinamine
5-Bromo-N,N-diethyl-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂
MDL Number: MFCD13561789
CAS Number: 1220027-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR