5-Bromo-4-methyl-2-(1-pyrrolidinyl)pyridine

Supplier: Matrix Scientific

051774-500MG 051774-2.5G
101896-988EA 364.62 USD
101896-988 101896-990
5-Bromo-4-methyl-2-(1-pyrrolidinyl)pyridine
5-Bromo-4-methyl-2-(1-pyrrolidinyl)pyridine

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂
MDL Number: MFCD12546611
CAS Number: 1187385-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR