5-Bromo-N,N,4-trimethyl-2-pyridinamine

Supplier: Matrix Scientific

051773-500MG 051773-2.5G
101896-984EA 301.54 USD
101896-984 101896-986
5-Bromo-N,N,4-trimethyl-2-pyridinamine
5-Bromo-N,N,4-trimethyl-2-pyridinamine

Hazard: Irritant

Formula: C₈H₁₁BrN₂
MDL Number: MFCD08272097
CAS Number: 764651-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR