4-[Methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]-3-nitrobenzoic acid

Supplier: Matrix Scientific

051772-500MG 051772-2.5G
101896-980EA 364.62 USD
101896-980 101896-982
4-[Methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]-3-nitrobenzoic acid
4-[Methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₅
MDL Number: MFCD13561788
CAS Number: 1219967-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR