4-[3,4-Dihydro-1(2H)-quinolinyl]-3-nitrobenzoic acid
Supplier: Matrix Scientific
051770-500MG
051770-2.5G
101896-972EA
364.62
USD
101896-972
101896-974
4-[3,4-Dihydro-1(2H)-quinolinyl]-3-nitrobenzoic acid
4-[3,4-Dihydro-1(2H)-quinolinyl]-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₆H₁₄N₂O₄ |
MDL Number:
MFCD13561786 CAS Number: 1220027-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...