4-(2,3-Dihydro-1H-indol-1-yl)-3-nitrobenzoic acid

Supplier: Matrix Scientific

051769-500MG 051769-2.5G
101896-968EA 364.62 USD
101896-968 101896-970
4-(2,3-Dihydro-1H-indol-1-yl)-3-nitrobenzoic acid
4-(2,3-Dihydro-1H-indol-1-yl)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₄
MDL Number: MFCD02937558
CAS Number: 451474-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR