4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-nitrobenzoic acid

Supplier: Matrix Scientific

051768-500MG 051768-2.5G
101896-964EA 364.62 USD
101896-964 101896-966
4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-nitrobenzoic acid
4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₄
MDL Number: MFCD02937562
CAS Number: 440347-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR