4-[4-(2-Hydroxyethyl)-1-piperazinyl]-3-nitrobenzoic acid

Supplier: Matrix Scientific

051767-500MG 051767-2.5G
101896-960EA 364.62 USD
101896-960 101896-962
4-[4-(2-Hydroxyethyl)-1-piperazinyl]-3-nitrobenzoic acid
4-[4-(2-Hydroxyethyl)-1-piperazinyl]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇N₃O₅
MDL Number: MFCD01461081
CAS Number: 337312-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR