4-[Cyclohexyl(ethyl)amino]-3-nitrobenzoic acid
Supplier: Matrix Scientific
051766-500MG
051766-2.5G
101896-956EA
364.62
USD
101896-956
101896-958
4-[Cyclohexyl(ethyl)amino]-3-nitrobenzoic acid
4-[Cyclohexyl(ethyl)amino]-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₅H₂₀N₂O₄ |
MDL Number:
MFCD13352576 CAS Number: 1036476-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...