4-[Benzyl(methyl)amino]-3-nitrobenzoic acid

Supplier: Matrix Scientific

051765-500MG 051765-2.5G
101896-952EA 364.62 USD
101896-952 101896-954
4-[Benzyl(methyl)amino]-3-nitrobenzoic acid
4-[Benzyl(methyl)amino]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₄
MDL Number: MFCD02652944
CAS Number: 361157-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR