4-(2-Ethyl-1-piperidinyl)-3-nitrobenzoic acid

Supplier: Matrix Scientific

051764-500MG 051764-2.5G
101896-948EA 364.62 USD
101896-948 101896-950
4-(2-Ethyl-1-piperidinyl)-3-nitrobenzoic acid
4-(2-Ethyl-1-piperidinyl)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₄
MDL Number: MFCD11039343
CAS Number: 1094563-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR