3-Nitro-4-(3-oxo-1-piperazinyl)benzoic acid

Supplier: Matrix Scientific

051761-500MG 051761-2.5G
101896-936EA 364.62 USD
101896-936 101896-938
3-Nitro-4-(3-oxo-1-piperazinyl)benzoic acid
3-Nitro-4-(3-oxo-1-piperazinyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₁N₃O₅
MDL Number: MFCD12445745
CAS Number: 1019359-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR