4-(4-Benzyl-1-piperazinyl)-3-nitrobenzoic acid

Supplier: Matrix Scientific

051760-500MG 051760-2.5G
101896-932EA 364.62 USD
101896-932 101896-934
4-(4-Benzyl-1-piperazinyl)-3-nitrobenzoic acid
4-(4-Benzyl-1-piperazinyl)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₈H₁₉N₃O₄
MDL Number: MFCD02089271
CAS Number: 312921-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR