4-(2-Methyl-1-piperidinyl)-3-nitrobenzoic acid

Supplier: Matrix Scientific

051759-500MG 051759-2.5G
101896-928EA 364.62 USD
101896-928 101896-930
4-(2-Methyl-1-piperidinyl)-3-nitrobenzoic acid
4-(2-Methyl-1-piperidinyl)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₄
MDL Number: MFCD11039342
CAS Number: 1019461-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR