4-[Ethyl(2-hydroxyethyl)amino]-3-nitrobenzoic acid

Supplier: Matrix Scientific

051757-500MG 051757-2.5G
101896-920EA 364.62 USD
101896-920 101896-922
4-[Ethyl(2-hydroxyethyl)amino]-3-nitrobenzoic acid
4-[Ethyl(2-hydroxyethyl)amino]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₅
MDL Number: MFCD13236885
CAS Number: 1179291-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR