4-[Butyl(methyl)amino]-3-nitrobenzoic acid

Supplier: Matrix Scientific

051756-500MG 051756-2.5G
101896-916EA 364.62 USD
101896-916 101896-918
4-[Butyl(methyl)amino]-3-nitrobenzoic acid
4-[Butyl(methyl)amino]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
MDL Number: MFCD11523578
CAS Number: 1019465-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR