4-(3-Hydroxy-1-pyrrolidinyl)-3-nitrobenzoic acid

Supplier: Matrix Scientific

051755-500MG 051755-2.5G
101896-912EA 364.62 USD
101896-912 101896-914
4-(3-Hydroxy-1-pyrrolidinyl)-3-nitrobenzoic acid
4-(3-Hydroxy-1-pyrrolidinyl)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₅
MDL Number: MFCD11038203
CAS Number: 1220027-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR