Ethyl-3-amino-4-[methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]benzoate
Supplier: Matrix Scientific
051750-500MG
051750-2.5G
101896-892EA
364.62
USD
101896-892
101896-894
Ethyl-3-amino-4-[methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]benzoate
Ethyl-3-amino-4-[methyl(tetrahydro-2H-pyran-4-ylmethyl)amino]benzoate
Hazard: Irritant
Formula:
C₁₆H₂₄N₂O₃ |
MDL Number:
MFCD13561785 CAS Number: 1219967-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...