Ethyl-3-amino-4-[benzyl(ethyl)amino]benzoate

Supplier: Matrix Scientific

051749-500MG 051749-2.5G
101896-888EA 364.62 USD
101896-888 101896-890
Ethyl-3-amino-4-[benzyl(ethyl)amino]benzoate
Ethyl-3-amino-4-[benzyl(ethyl)amino]benzoate

Hazard: Irritant

Formula: C₁₈H₂₂N₂O₂
MDL Number: MFCD13561784
CAS Number: 1220036-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR