Ethyl-3-amino-4-[3,4-dihydro-1(2H)-quinolinyl]benzoate

Supplier: Matrix Scientific

051748-500MG 051748-2.5G
101896-884EA 364.62 USD
101896-884 101896-886
Ethyl-3-amino-4-[3,4-dihydro-1(2H)-quinolinyl]benzoate
Ethyl-3-amino-4-[3,4-dihydro-1(2H)-quinolinyl]benzoate

Hazard: Irritant

Formula: C₁₈H₂₀N₂O₂
MDL Number: MFCD13561783
CAS Number: 1220019-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR