Ethyl-3-amino-4-[3,4-dihydro-2(1H)-isoquinolinyl]benzoate

Supplier: Matrix Scientific

051746-500MG 051746-2.5G
101896-876EA 364.62 USD
101896-876 101896-878
Ethyl-3-amino-4-[3,4-dihydro-2(1H)-isoquinolinyl]benzoate
Ethyl-3-amino-4-[3,4-dihydro-2(1H)-isoquinolinyl]benzoate

Hazard: Irritant

Formula: C₁₈H₂₀N₂O₂
MDL Number: MFCD13561781
CAS Number: 1220036-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR