Ethyl-3-amino-4-(4-hydroxy-1-piperidinyl)benzoate

Supplier: Matrix Scientific

051739-500MG 051739-2.5G
101896-848EA 364.62 USD
101896-848 101896-850
Ethyl-3-amino-4-(4-hydroxy-1-piperidinyl)benzoate
Ethyl-3-amino-4-(4-hydroxy-1-piperidinyl)benzoate

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₃
MDL Number: MFCD13344437
CAS Number: 1219967-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR