Ethyl-3-amino-4-(3-oxo-1-piperazinyl)benzoate

Supplier: Matrix Scientific

051738-500MG 051738-2.5G
101896-844EA 364.62 USD
101896-844 101896-846
Ethyl-3-amino-4-(3-oxo-1-piperazinyl)benzoate
Ethyl-3-amino-4-(3-oxo-1-piperazinyl)benzoate

Hazard: Irritant

Formula: C₁₃H₁₇N₃O₃
MDL Number: MFCD13344472
CAS Number: 1219964-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR