Ethyl-3-amino-4-(1-azepanyl)benzoate
Supplier: Matrix Scientific
051737-500MG
051737-2.5G
101896-840EA
364.62
USD
101896-840
101896-842
Ethyl-3-amino-4-(1-azepanyl)benzoate
Ethyl-3-amino-4-(1-azepanyl)benzoate
Hazard: Irritant
Formula:
C₁₅H₂₂N₂O₂ |
MDL Number:
MFCD13343706 CAS Number: 343617-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...