Ethyl-3-amino-4-(4-benzyl-1-piperazinyl)benzoate

Supplier: Matrix Scientific

Synonyms: DCLX069, Ethyl 3-amino-4-(4-benzylpiperazin-1-yl)benzoate

051736-500MG 051736-2.5G
101896-836EA 364.62 USD
101896-836 101896-838
Ethyl-3-amino-4-(4-benzyl-1-piperazinyl)benzoate
Ethyl-3-amino-4-(4-benzyl-1-piperazinyl)benzoate

Hazard: Irritant

Formula: C₂₀H₂₅N₃O₂
MW: 339.44 g/mol
MDL Number: MFCD06666140
CAS Number: 792946-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR