Ethyl-3-amino-4-(4-methyl-1-piperazinyl)benzoate

Supplier: Matrix Scientific

051735-500MG 051735-2.5G
101896-832EA 364.62 USD
101896-832 101896-834
Ethyl-3-amino-4-(4-methyl-1-piperazinyl)benzoate
Ethyl-3-amino-4-(4-methyl-1-piperazinyl)benzoate

Hazard: Irritant

Formula: C₁₄H₂₁N₃O₂
MDL Number: MFCD13343616
CAS Number: 1220036-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR