Ethyl-3-amino-4-(4-methyl-1-piperidinyl)benzoate

Supplier: Matrix Scientific

051734-500MG 051734-2.5G
101896-828EA 364.62 USD
101896-828 101896-830
Ethyl-3-amino-4-(4-methyl-1-piperidinyl)benzoate
Ethyl-3-amino-4-(4-methyl-1-piperidinyl)benzoate

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD13343546
CAS Number: 1220019-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR