Ethyl-3-amino-4-(3-methyl-1-piperidinyl)benzoate

Supplier: Matrix Scientific

051733-500MG 051733-2.5G
101896-824EA 364.62 USD
101896-824 101896-826
Ethyl-3-amino-4-(3-methyl-1-piperidinyl)benzoate
Ethyl-3-amino-4-(3-methyl-1-piperidinyl)benzoate

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD13343697
CAS Number: 1219960-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR