Ethyl-3-amino-4-(2-methyl-1-piperidinyl)benzoate

Supplier: Matrix Scientific

051732-500MG 051732-2.5G
101896-820EA 364.62 USD
101896-820 101896-822
Ethyl-3-amino-4-(2-methyl-1-piperidinyl)benzoate
Ethyl-3-amino-4-(2-methyl-1-piperidinyl)benzoate

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD13343595
CAS Number: 1141473-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR