Ethyl-3-amino-4-(diallylamino)benzoate
Supplier: Matrix Scientific
051731-500MG
051731-2.5G
101896-816EA
364.62
USD
101896-816
101896-818
Ethyl-3-amino-4-(diallylamino)benzoate
Ethyl-3-amino-4-(diallylamino)benzoate
Hazard: Irritant
Formula:
C₁₅H₂₀N₂O₂ |
MDL Number:
MFCD13561775 CAS Number: 1219967-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...