Ethyl-3-amino-4-(3-hydroxy-1-pyrrolidinyl)benzoate

Supplier: Matrix Scientific

051728-500MG 051728-2.5G
101896-804EA 364.62 USD
101896-804 101896-806
Ethyl-3-amino-4-(3-hydroxy-1-pyrrolidinyl)benzoate
Ethyl-3-amino-4-(3-hydroxy-1-pyrrolidinyl)benzoate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD13561774
CAS Number: 1219967-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR