Ethyl-3-amino-4-(4-morpholinyl)benzoate
Supplier: Matrix Scientific
051727-500MG
051727-2.5G
101896-800EA
364.62
USD
101896-800
101896-802
Ethyl-3-amino-4-(4-morpholinyl)benzoate
Ethyl-3-amino-4-(4-morpholinyl)benzoate
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₃ |
MDL Number:
MFCD00277379 CAS Number: 71254-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...