Ethyl-3-amino-4-(1-piperidinyl)benzoate

Supplier: Matrix Scientific

051726-500MG 051726-2.5G
101896-796EA 364.62 USD
101896-796 101896-798
Ethyl-3-amino-4-(1-piperidinyl)benzoate
Ethyl-3-amino-4-(1-piperidinyl)benzoate

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MDL Number: MFCD00238972
CAS Number: 71254-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR