Ethyl-3-amino-4-[(2-hydroxyethyl)(methyl)amino]benzoate

Supplier: Matrix Scientific

051725-500MG 051725-2.5G
101896-792EA 364.62 USD
101896-792 101896-794
Ethyl-3-amino-4-[(2-hydroxyethyl)(methyl)amino]benzoate
Ethyl-3-amino-4-[(2-hydroxyethyl)(methyl)amino]benzoate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃
MDL Number: MFCD13344939
CAS Number: 1219964-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR