Ethyl-3-amino-4-(diethylamino)benzoate

Supplier: Matrix Scientific

051724-500MG 051724-2.5G
101896-788EA 364.62 USD
101896-788 101896-790
Ethyl-3-amino-4-(diethylamino)benzoate
Ethyl-3-amino-4-(diethylamino)benzoate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MDL Number: MFCD13343588
CAS Number: 1220027-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR