Ethyl-3-amino-4-(1-pyrrolidinyl)benzoate

Supplier: Matrix Scientific

051723-500MG 051723-2.5G
101896-784EA 364.62 USD
101896-784 101896-786
Ethyl-3-amino-4-(1-pyrrolidinyl)benzoate
Ethyl-3-amino-4-(1-pyrrolidinyl)benzoate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD13343551
CAS Number: 343594-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR