Ethyl-3-amino-4-(dimethylamino)benzoate

Supplier: Matrix Scientific

051722-500MG 051722-2.5G
101896-780EA 364.62 USD
101896-780 101896-782
Ethyl-3-amino-4-(dimethylamino)benzoate
Ethyl-3-amino-4-(dimethylamino)benzoate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD11556389
CAS Number: 178469-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR