N-Methyl-N-(2-pyrrolidinylmethyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)amine dihydrochloride
Supplier: Matrix Scientific
051721-500MG
051721-2.5G
101896-776EA
364.62
USD
101896-776
101896-778
N-Methyl-N-(2-pyrrolidinylmethyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)amine dihydrochloride
N-Methyl-N-(2-pyrrolidinylmethyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)amine dihydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₆Cl₂N₂O |
MDL Number:
MFCD13561773 CAS Number: 1220027-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...