N-Benzyl-N-(2-pyrrolidinylmethyl)-1-ethanamine dihydrochloride

Supplier: Matrix Scientific

051720-500MG 051720-2.5G
101896-772EA 364.62 USD
101896-772 101896-774
N-Benzyl-N-(2-pyrrolidinylmethyl)-1-ethanamine dihydrochloride
N-Benzyl-N-(2-pyrrolidinylmethyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₄Cl₂N₂
MW: 291.27 g/mol
MDL Number: MFCD13561772
CAS Number: 1220036-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR