1-(2-Pyrrolidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Supplier: Matrix Scientific

051719-500MG 051719-2.5G
101896-768EA 364.62 USD
101896-768 101896-770
1-(2-Pyrrolidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride
1-(2-Pyrrolidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂Cl₂N₂
MDL Number: MFCD13561771
CAS Number: 1220027-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR