2-(2-Pyrrolidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Supplier: Matrix Scientific

051717-500MG 051717-2.5G
101896-760EA 364.62 USD
101896-760 101896-762
2-(2-Pyrrolidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
2-(2-Pyrrolidinylmethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂Cl₂N₂
MW: 289.25 g/mol
MDL Number: MFCD13561769
CAS Number: 1220019-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR