2-[4-(2-Pyrrolidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051716-500MG 051716-2.5G
101896-756EA 166.15 USD
101896-756 101896-758
2-[4-(2-Pyrrolidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride
2-[4-(2-Pyrrolidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅Cl₂N₃O
MW: 286.25 g/mol
MDL Number: MFCD13561768
CAS Number: 1220027-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR