N-Ethyl-N-(2-pyrrolidinylmethyl)cyclohexanamine dihydrochloride

Supplier: Matrix Scientific

051715-500MG 051715-2.5G
101896-752EA 364.62 USD
101896-752 101896-754
N-Ethyl-N-(2-pyrrolidinylmethyl)cyclohexanamine dihydrochloride
N-Ethyl-N-(2-pyrrolidinylmethyl)cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561767
CAS Number: 1219964-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR