N-Methyl(phenyl)-N-(2-pyrrolidinylmethyl)methanamine dihydrochloride

Supplier: Matrix Scientific

051714-500MG 051714-2.5G
101896-748EA 364.62 USD
101896-748 101896-750
N-Methyl(phenyl)-N-(2-pyrrolidinylmethyl)methanamine dihydrochloride
N-Methyl(phenyl)-N-(2-pyrrolidinylmethyl)methanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₂Cl₂N₂
MW: 277.24 g/mol
MDL Number: MFCD13561766
CAS Number: 1220036-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR