2-Ethyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride
Supplier: Matrix Scientific
051713-500MG
051713-2.5G
101896-744EA
364.62
USD
101896-744
101896-746
2-Ethyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride
2-Ethyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₆Cl₂N₂ MW: 269.26 g/mol |
MDL Number:
MFCD13561765 CAS Number: 1220027-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...