N-Propyl-N-(2-pyrrolidinylmethyl)-1-propanamine dihydrochloride

Supplier: Matrix Scientific

051711-500MG 051711-2.5G
101896-736EA 364.62 USD
101896-736 101896-738
N-Propyl-N-(2-pyrrolidinylmethyl)-1-propanamine dihydrochloride
N-Propyl-N-(2-pyrrolidinylmethyl)-1-propanamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₆Cl₂N₂
MDL Number: MFCD13561763
CAS Number: 1220035-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR