1-(2-Pyrrolidinylmethyl)-4-piperidinol dihydrochloride

Supplier: Matrix Scientific

051710-500MG 051710-2.5G
101896-732EA 364.62 USD
101896-732 101896-734
1-(2-Pyrrolidinylmethyl)-4-piperidinol dihydrochloride
1-(2-Pyrrolidinylmethyl)-4-piperidinol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂O
MDL Number: MFCD13561762
CAS Number: 1219964-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR