4-(2-Pyrrolidinylmethyl)-2-piperazinone dihydrochloride

Supplier: Matrix Scientific

051709-500MG 051709-2.5G
101896-728EA 364.62 USD
101896-728 101896-730
4-(2-Pyrrolidinylmethyl)-2-piperazinone dihydrochloride
4-(2-Pyrrolidinylmethyl)-2-piperazinone dihydrochloride

Hazard: Irritant

Formula: C₉H₁₉Cl₂N₃O
MDL Number: MFCD13561761
CAS Number: 1220027-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR