1-(2-Pyrrolidinylmethyl)azepane dihydrochloride
Supplier: Matrix Scientific
051708-500MG
051708-2.5G
101896-724EA
364.62
USD
101896-724
101896-726
1-(2-Pyrrolidinylmethyl)azepane dihydrochloride
1-(2-Pyrrolidinylmethyl)azepane dihydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₄Cl₂N₂ MW: 255.23 g/mol |
MDL Number:
MFCD13561760 CAS Number: 1219964-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...