1-Benzyl-4-(2-pyrrolidinylmethyl)piperazine dihydrochloride

Supplier: Matrix Scientific

051707-500MG 051707-2.5G
101896-720EA 364.62 USD
101896-720 101896-722
1-Benzyl-4-(2-pyrrolidinylmethyl)piperazine dihydrochloride
1-Benzyl-4-(2-pyrrolidinylmethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₆H₂₇Cl₂N₃
MDL Number: MFCD13561759
CAS Number: 1220036-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR